Learn More
Medchemexpress LLC Sultopride hydrochloride | 23694-17-9 | 99.9% | 390.93 | 50 MG
SDP
Catalog No. 5000405094
Encompass_Preferred
Sultopride hydrochloride is a selective antagonist of the dopamine D2 receptor. It has shown increased levels of DOPAC and HVA in the striatum, nucleus accumbens, and medial prefrontal cortex in treated rats.
  • Acts as a selective antagonist of dopamine D2 receptor.
  • Demonstrates high purity of 99.92%.
  • Soluble in water and DMSO for in vitro applications.
  • Suitable for in vivo studies with protocol for PBS solubility.
  • Available in solid form, white to off-white in appearance.
  • Stored at 4°C, sealed, away from moisture.
Catalog No. 50-004-05094 Supplier Medchemexpress LLC Supplier No. HY42849A50MG
Sultopride hydrochloride is a selective antagonist of the dopamine D2 receptor. It has shown increased levels of DOPAC and HVA in the striatum, nucleus accumbens, and medial prefrontal cortex in treated rats.
  • Acts as a selective antagonist of dopamine D2 receptor.
  • Demonstrates high purity of 99.92%.
  • Soluble in water and DMSO for in vitro applications.
  • Suitable for in vivo studies with protocol for PBS solubility.
  • Available in solid form, white to off-white in appearance.
  • Stored at 4°C, sealed, away from moisture.