Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sultopride hydrochloride | 23694-17-9 | MFCD01319147 | 99.9% | 390.93 | C17H27ClN2O4S | 10 MG
SDP

Catalog No. 5000299247
Encompass_Preferred

Sultopride hydrochloride (LIN-1418 hydrochloride) is a selective dopamine D2 receptor antagonist supplied as a white to off-white solid for laboratory research applications. It is provided at high reported purity and in small pack sizes for bench-top experiments.

  • Selective dopamine D2 receptor antagonist.
  • High purity (≈99.9%).
  • White to off-white solid physical form.
  • CAS number 23694-17-9.
  • Molecular weight 390.93 g/mol.
  • Available in small pack sizes, including 10 MG.
  • Intended for research use only; not for human or clinical use.
  • Storage: 4°C, sealed and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-99247 Supplier Medchemexpress LLC Supplier No. HY42849A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sultopride hydrochloride (LIN-1418 hydrochloride) is a selective dopamine D2 receptor antagonist supplied as a white to off-white solid for laboratory research applications. It is provided at high reported purity and in small pack sizes for bench-top experiments.

  • Selective dopamine D2 receptor antagonist.
  • High purity (≈99.9%).
  • White to off-white solid physical form.
  • CAS number 23694-17-9.
  • Molecular weight 390.93 g/mol.
  • Available in small pack sizes, including 10 MG.
  • Intended for research use only; not for human or clinical use.
  • Storage: 4°C, sealed and away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.