Learn More
Medchemexpress LLC Sultopride hydrochloride (LIN-1418 hydrochloride) | 23694-17-9 | 99.9% | 390.93 | 100 MG
SDP
Catalog No. 5000405093
Encompass_Preferred
Sultopride hydrochloride is a selective antagonist of dopamine D2 receptor, suitable for research use only.
  • Functions as a selective dopamine D2 receptor antagonist.
  • Increases DOPAC and HVA levels in the striatum, nucleus accumbens, and medial prefrontal cortex in rats.
  • Demonstrates higher DOPAC and HVA levels in treated rats compared to sulpiride-treated rats in the striatum and nucleus accumbens.
Catalog No. 50-004-05093 Supplier Medchemexpress LLC Supplier No. HY42849A100MG
Sultopride hydrochloride is a selective antagonist of dopamine D2 receptor, suitable for research use only.
  • Functions as a selective dopamine D2 receptor antagonist.
  • Increases DOPAC and HVA levels in the striatum, nucleus accumbens, and medial prefrontal cortex in rats.
  • Demonstrates higher DOPAC and HVA levels in treated rats compared to sulpiride-treated rats in the striatum and nucleus accumbens.