Learn More
Medchemexpress LLC Sultosilic acid piperazine salt | 57775-27-6 | 25 MG
SDP
Catalog No. 5000341447
Encompass_Preferred
Sultosilic acid piperazine salt is a lipid-lowering agent that modifies blood lipid levels and reduces platelet adhesiveness without promoting peroxisomal activity of hepatocytes or causing other adverse side-effects. It is for research use only.
  • Modifies blood lipid levels
  • Reduces platelet adhesiveness
  • Does not promote peroxisomal activity of hepatocytes
  • Does not cause other adverse side-effects
Catalog No. 50-003-41447 Supplier Medchemexpress LLC Supplier No. 25MGHY130587
Sultosilic acid piperazine salt is a lipid-lowering agent that modifies blood lipid levels and reduces platelet adhesiveness without promoting peroxisomal activity of hepatocytes or causing other adverse side-effects. It is for research use only.
  • Modifies blood lipid levels
  • Reduces platelet adhesiveness
  • Does not promote peroxisomal activity of hepatocytes
  • Does not cause other adverse side-effects