Learn More
TARGETMOL CHEMICALS INC Sultosilic acid piperazine
SDP
Catalog No. 502877672
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sultosilic acid piperazine salt (A-585) is a novel lipid-lowering compound that can be used to study type IIb familial combined hyperlipidemia. Purity 99.1%
Catalog No. 50-287-7672 Supplier TARGETMOL CHEMICALS INC Supplier No. T288821MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Sultosilic acid piperazine salt (A-585) is a novel lipid-lowering compound that can be used to study type IIb familial combined hyperlipidemia. Purity 99.1%