Learn More
MSC Diversey Presoak Destainer, 1 gal, Alkaline-Chlorinated, China/Stainless Steel, 4/CT
Catalog No. 502815522
Encompass
Suma Dip K1 is an alkaline-chlorinated liquid presoak and destainer specifically formulated for the removal of stubborn stains and soils from cups, china, and stainless steel. This powerful formula contains scale inhibitors, caustic alkali, and active chlorine to effectively remove tannin, tea, coffee, and fruit stains, as well as burnt-on food residues and starch.
  • Removes stubborn tannin, tea, coffee, and fruit stains
  • Effectively assists in the removal of burnt-on food residues and starch
  • Easy to dose manually or with a dosing pump
  • Formulated for use on china and stainless steel
  • Concentrate with a dilution rate of 1:32
Catalog No. 50-281-5522 Supplier MSC Supplier No. 03108859
Suma Dip K1 is an alkaline-chlorinated liquid presoak and destainer specifically formulated for the removal of stubborn stains and soils from cups, china, and stainless steel. This powerful formula contains scale inhibitors, caustic alkali, and active chlorine to effectively remove tannin, tea, coffee, and fruit stains, as well as burnt-on food residues and starch.
  • Removes stubborn tannin, tea, coffee, and fruit stains
  • Effectively assists in the removal of burnt-on food residues and starch
  • Easy to dose manually or with a dosing pump
  • Formulated for use on china and stainless steel
  • Concentrate with a dilution rate of 1:32