Learn More
MSC Diversey Drain Maintainer, Liquid, Unscented, 5 gal
Catalog No. 502817288
Encompass
This liquid drain maintainer is designed for commercial and industrial use, effectively extending manual cleaning cycles and restoring normal flow in slow drains. Its highly concentrated, non-toxic, non-corrosive, and non-caustic formula ensures safe and economical application.
  • Significantly extends the manual cleaning cycle
  • Restores normal flow rate to slow drains
  • Reduces the demand for further downstream waste processing
  • Non-toxic, non-corrosive, non-caustic, and safe for general commercial and industrial use
  • Highly concentrated formula gives greater economy in use
  • Unscented liquid form
  • Safe for use in grease traps, septic systems, drains, and pipes
Catalog No. 50-281-7288 Supplier MSC Supplier No. 02942787
This liquid drain maintainer is designed for commercial and industrial use, effectively extending manual cleaning cycles and restoring normal flow in slow drains. Its highly concentrated, non-toxic, non-corrosive, and non-caustic formula ensures safe and economical application.
  • Significantly extends the manual cleaning cycle
  • Restores normal flow rate to slow drains
  • Reduces the demand for further downstream waste processing
  • Non-toxic, non-corrosive, non-caustic, and safe for general commercial and industrial use
  • Highly concentrated formula gives greater economy in use
  • Unscented liquid form
  • Safe for use in grease traps, septic systems, drains, and pipes