Learn More
MSC Diversey Cleaner / Deodorizer, Floral Scent, Liquid, 32 oz
Catalog No. 502838608
Encompass
This Suma Dumpster Fresh D2.2 Cleaner / Deodorizer is a green, liquid solution designed to eliminate unbearable odors and clean organic waste from dumpsters, trash cans, loading docks, and concrete floors. Its bio-active formula quickly works to create enzymes that digest grease and other organic soils, offering a comprehensive cleaning and deodorizing solution.
  • Eliminates unbearable odors quickly and easily.
  • Cleans away organic waste that attracts rodents, insects, and other pests.
  • Requires no scrubbing.
  • Applied bacteria keep odors from coming back and clean the soil that's hard to reach.
  • Use to clean and deodorize dumpsters, trash cans, loading docks, and concrete floors.
  • For trash cans, apply product 3 times per week when cans are emptied.
  • For dumpsters and surrounding area, apply product when dumpster is emptied.
Catalog No. 50-283-8608 Supplier MSC Supplier No. 50637149
This Suma Dumpster Fresh D2.2 Cleaner / Deodorizer is a green, liquid solution designed to eliminate unbearable odors and clean organic waste from dumpsters, trash cans, loading docks, and concrete floors. Its bio-active formula quickly works to create enzymes that digest grease and other organic soils, offering a comprehensive cleaning and deodorizing solution.
  • Eliminates unbearable odors quickly and easily.
  • Cleans away organic waste that attracts rodents, insects, and other pests.
  • Requires no scrubbing.
  • Applied bacteria keep odors from coming back and clean the soil that's hard to reach.
  • Use to clean and deodorize dumpsters, trash cans, loading docks, and concrete floors.
  • For trash cans, apply product 3 times per week when cans are emptied.
  • For dumpsters and surrounding area, apply product when dumpster is emptied.