Learn More
MSC Diversey Floor Cleaner, Liquid, Cold Room, 4 ea
Catalog No. 502867210
Encompass
Suma Freeze D2.9 is a ready-to-use floor cleaner designed for effective cleaning in extreme conditions, including temperatures as low as -29° C. This cleaner facilitates cold room and freezer floor maintenance without the need for defrosting, leaving surfaces clean and residue-free.
  • Works effectively in temperatures as low as -29° C
  • Cleans cold room and freezer floors without defrosting
  • Leaves floors clean with no residue
  • Phosphate-free formula
  • Versatile and easy to use
Catalog No. 50-286-7210 Supplier MSC Supplier No. 20465902
Suma Freeze D2.9 is a ready-to-use floor cleaner designed for effective cleaning in extreme conditions, including temperatures as low as -29° C. This cleaner facilitates cold room and freezer floor maintenance without the need for defrosting, leaving surfaces clean and residue-free.
  • Works effectively in temperatures as low as -29° C
  • Cleans cold room and freezer floors without defrosting
  • Leaves floors clean with no residue
  • Phosphate-free formula
  • Versatile and easy to use