Learn More
MSC Diversey Stainless Steel Polish, Oil-Based, Unscented, 32 oz, 6/Pk.
Catalog No. 502815467
Encompass
Suma Inox D7 is an oil-based, ready-to-use cleaner and polish for stainless steel non-food contact surfaces and other metal surfaces like brass, aluminum, and copper. It is ideal for use on refrigerator doors, handles, trolleys, and dishwashing machines, leaving surfaces clean and shiny.
  • Helps protect surfaces by forming a protective film
  • Resists finger marks and water spotting to maintain a polished appearance for longer
  • Keeps surfaces looking good, with a smooth luster
Catalog No. 50-281-5467 Supplier MSC Supplier No. 20467361
Suma Inox D7 is an oil-based, ready-to-use cleaner and polish for stainless steel non-food contact surfaces and other metal surfaces like brass, aluminum, and copper. It is ideal for use on refrigerator doors, handles, trolleys, and dishwashing machines, leaving surfaces clean and shiny.
  • Helps protect surfaces by forming a protective film
  • Resists finger marks and water spotting to maintain a polished appearance for longer
  • Keeps surfaces looking good, with a smooth luster