Learn More
MSC Diversey Suma Rinse A5, Economy, 1 gal, 4/Carton
Catalog No. 502817299
Encompass
Suma® Rinse A5 is an economy rinse aid designed for soft-to-medium water conditions. It is specifically formulated for use in a wide range of warewashing machines. The product contains a special blend of nonionic surfactants to facilitate rapid spot and streak-free drying.
  • Low foaming action for more efficient operation
  • Gives streak and spot free results
  • Ensures quick drying
  • Low foam general purpose rinse additive for machine warewashing
Catalog No. 50-281-7299 Supplier MSC Supplier No. 02964823
Suma® Rinse A5 is an economy rinse aid designed for soft-to-medium water conditions. It is specifically formulated for use in a wide range of warewashing machines. The product contains a special blend of nonionic surfactants to facilitate rapid spot and streak-free drying.
  • Low foaming action for more efficient operation
  • Gives streak and spot free results
  • Ensures quick drying
  • Low foam general purpose rinse additive for machine warewashing