Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Suma Pan-Clean

Catalog No. 17000053
Change view
Click to view available options
Color:
Dark Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-000-053 Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-000-053 Supplier Diversey™ Supplier No. 903836
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly concentrated dish detergent

  • Quickly removes grease and food residue from soiled areas
  • Long-lasting suds provide outstanding cleaning performance
  • Delivers superior grease cutting on toughest soils
  • Specially-formulated to leave sink areas smelling clean and fresh
  • Non-corrosive and high concentrated for good cost in use
  • Leaves equipment and utensils thoroughly clean
  • AccuMix™ D1.5
  • Ratio: 1:256, 1:512
  • Floral scent

Specifications

Product Type Suma-Pan Clean
Sterility Non-sterile
Appearance Dark Blue
Certifications/Compliance Kosher, Halal, Letter of no objection
Color Dark Blue
Filtered Non-filtered
For Use With (Application) For handwashing of pots, pans, utensils, glassware and other food department equipment, Cleaning three-compartment sinks
Quantity 6 Pk.
Volume (English) 32 oz.
Volume (Metric) 946 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.