Learn More
MSC Diversey Suma Stop Slip Traction Treatment, Dilutable Concentrate, 1 gal
Catalog No. 502819610
Encompass
Diversey Suma Stop Slip Traction Treatment is a dilutable concentrate formulated with sodium sulfonates, designed for application on various floor surfaces including ceramic tile, glazed tile, marble, porcelain, and terrazzo to improve traction.
  • Container type: bottle
  • Container size: 1.00 gal
  • Solution type: dilutable concentrate
  • Material application: porcelain, glazed tile, ceramic tiles, terrazzo, marble
  • Formula type: sodium sulfonates
Catalog No. 50-281-9610 Supplier MSC Supplier No. 20466256
Diversey Suma Stop Slip Traction Treatment is a dilutable concentrate formulated with sodium sulfonates, designed for application on various floor surfaces including ceramic tile, glazed tile, marble, porcelain, and terrazzo to improve traction.
  • Container type: bottle
  • Container size: 1.00 gal
  • Solution type: dilutable concentrate
  • Material application: porcelain, glazed tile, ceramic tiles, terrazzo, marble
  • Formula type: sodium sulfonates