Learn More
MSC Solenis Suma Ultra L2 Mechanical Detergent, Non-Chlorinated, 1 gal, 4/Carton
Catalog No. 502817121
Encompass
Suma Ultra L2 Mechanical Warewashing Detergent is a non-chlorinated machine dish liquid engineered for superior soil removal in soft water environments. It contains scale control agents and is designed for efficient, effective warewashing.
  • Cuts through grease and dried-on food
  • Prevents scaling of items and machine parts in soft water
  • Soil suspension agents prevent food and stains from redepositing onto ware
  • Highly concentrated formulation for greater economy in use and fewer container changes
  • Low foaming action for more efficient and effective operation
  • Auto-dosed
  • Ideal for soft water conditions
  • Not suitable for aluminum or other soft metals
Catalog No. 50-281-7121 Supplier MSC Supplier No. 03108560
Suma Ultra L2 Mechanical Warewashing Detergent is a non-chlorinated machine dish liquid engineered for superior soil removal in soft water environments. It contains scale control agents and is designed for efficient, effective warewashing.
  • Cuts through grease and dried-on food
  • Prevents scaling of items and machine parts in soft water
  • Soil suspension agents prevent food and stains from redepositing onto ware
  • Highly concentrated formulation for greater economy in use and fewer container changes
  • Low foaming action for more efficient and effective operation
  • Auto-dosed
  • Ideal for soft water conditions
  • Not suitable for aluminum or other soft metals