Learn More
Medchemexpress LLC Sumanirole (maleate) | 179386-44-8 | 99.8% | 319.31 | C15H17N3O5 | 10 MG
SDP
Catalog No. 5000298633
Encompass_Preferred
Sumanirole maleate is the maleate salt of sumanirole, a selective dopamine D2 receptor full agonist used in neuroscience and pharmacology research to investigate dopaminergic signaling and disease models. It is supplied as a white to off-white solid with high purity and specified molecular formula C15H17N3O5 (MW 319.31).
  • Selective D2 receptor full agonist for receptor pharmacology studies.
  • High purity suitable for analytical and biological assays.
  • White to off-white solid appearance for easy visual inspection.
  • Available in small research quantities appropriate for in vitro studies.
  • Stable when stored sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • CAS number 179386-44-8 for unambiguous chemical identification.
Catalog No. 50-002-98633 Supplier Medchemexpress LLC Supplier No. HY70081A10MG
Sumanirole maleate is the maleate salt of sumanirole, a selective dopamine D2 receptor full agonist used in neuroscience and pharmacology research to investigate dopaminergic signaling and disease models. It is supplied as a white to off-white solid with high purity and specified molecular formula C15H17N3O5 (MW 319.31).
  • Selective D2 receptor full agonist for receptor pharmacology studies.
  • High purity suitable for analytical and biological assays.
  • White to off-white solid appearance for easy visual inspection.
  • Available in small research quantities appropriate for in vitro studies.
  • Stable when stored sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • CAS number 179386-44-8 for unambiguous chemical identification.