Learn More
Medchemexpress LLC Sumanirole maleate | 179386-44-8 | 99.8% | C15H17N3O5 | 1 ML
SDP
Catalog No. 5000434219
Encompass_Preferred
Sumanirole maleate is a highly selective D2 receptor full agonist with an ED50 of approximately 46 nM. It is important in the research of Parkinson's disease and restless leg syndrome. Sumanirole serves as a valuable tool compound for basic research to identify neurobiological mechanisms based on a dopamine D2-linked mechanism of action.
  • Highly selective D2 receptor full agonist
  • ED50 of approximately 46 nM
  • Important in research of Parkinson's disease
  • Important in research of restless leg syndrome
  • Valuable tool compound for basic research
Catalog No. 50-004-34219 Supplier Medchemexpress LLC Supplier No. HY70081A10MM/1ML
Sumanirole maleate is a highly selective D2 receptor full agonist with an ED50 of approximately 46 nM. It is important in the research of Parkinson's disease and restless leg syndrome. Sumanirole serves as a valuable tool compound for basic research to identify neurobiological mechanisms based on a dopamine D2-linked mechanism of action.
  • Highly selective D2 receptor full agonist
  • ED50 of approximately 46 nM
  • Important in research of Parkinson's disease
  • Important in research of restless leg syndrome
  • Valuable tool compound for basic research