Learn More
Medchemexpress LLC Sumanirole (maleate) | 179386-44-8 | 99.8% | C15H17N3O5 | 25 MG
SDP
Catalog No. 5000358855
Encompass_Preferred
Sumanirole maleate (U-95666E; PNU-95666E) is a highly selective D2 receptor full agonist with an ED50 of about 46 nM. It plays an important role in the research of Parkinson's disease and restless leg syndrome. Sumanirole is a valuable tool compound for basic research to identify neurobiological mechanisms based on a dopamine D2-linked (vs. D1, D3, D4, and D5-linked) mechanism of action.
  • Highly selective D2 receptor full agonist
  • ED50 of about 46 nM
  • Valuable tool for identifying neurobiological mechanisms
  • Used in research for Parkinson's disease
  • Used in research for restless leg syndrome
Catalog No. 50-003-58855 Supplier Medchemexpress LLC Supplier No. 25MGHY70081A
Sumanirole maleate (U-95666E; PNU-95666E) is a highly selective D2 receptor full agonist with an ED50 of about 46 nM. It plays an important role in the research of Parkinson's disease and restless leg syndrome. Sumanirole is a valuable tool compound for basic research to identify neurobiological mechanisms based on a dopamine D2-linked (vs. D1, D3, D4, and D5-linked) mechanism of action.
  • Highly selective D2 receptor full agonist
  • ED50 of about 46 nM
  • Valuable tool for identifying neurobiological mechanisms
  • Used in research for Parkinson's disease
  • Used in research for restless leg syndrome