Learn More
Medchemexpress LLC Sumanirole maleate | 179386-44-8 | 99.8% | C15H17N3O5 | 50 MG
SDP
Catalog No. 5000434579
Encompass_Preferred
Sumanirole maleate is a highly selective full agonist of the D2 receptor, exhibiting an ED50 of approximately 46 nM. This compound is significant in the study of Parkinson's disease and restless leg syndrome. It serves as a valuable research tool for investigating neurobiological mechanisms that are specifically linked to dopamine D2 receptor activity.
  • Highly selective full agonist of the D2 receptor.
  • Involved in research concerning Parkinson's disease.
  • Used in the study of restless leg syndrome.
  • Aids in identifying neurobiological mechanisms dependent on D2 receptor linkage.
Catalog No. 50-004-34579 Supplier Medchemexpress LLC Supplier No. HY70081A50MG
Sumanirole maleate is a highly selective full agonist of the D2 receptor, exhibiting an ED50 of approximately 46 nM. This compound is significant in the study of Parkinson's disease and restless leg syndrome. It serves as a valuable research tool for investigating neurobiological mechanisms that are specifically linked to dopamine D2 receptor activity.
  • Highly selective full agonist of the D2 receptor.
  • Involved in research concerning Parkinson's disease.
  • Used in the study of restless leg syndrome.
  • Aids in identifying neurobiological mechanisms dependent on D2 receptor linkage.