Learn More
Medchemexpress LLC Sumanirole maleate (U-95666E; PNU-95666E) | 179386-44-8 | 99.8% | C15H17N3O5 | 100 MG
SDP
Catalog No. 5000433713
Encompass_Preferred
Sumanirole maleate (U-95666E; PNU-95666E) is a highly selective D2 receptor full agonist with an ED50 of about 46 nM. It plays an important role in the research of Parkinson's disease and restless leg syndrome.
  • Highly selective D2 receptor full agonist
  • ED50 of about 46 nM
  • Valuable tool compound for basic research
  • Identifies neurobiological mechanisms based on dopamine D2-linked mechanism of action
Catalog No. 50-004-33713 Supplier Medchemexpress LLC Supplier No. HY70081A100MG
Sumanirole maleate (U-95666E; PNU-95666E) is a highly selective D2 receptor full agonist with an ED50 of about 46 nM. It plays an important role in the research of Parkinson's disease and restless leg syndrome.
  • Highly selective D2 receptor full agonist
  • ED50 of about 46 nM
  • Valuable tool compound for basic research
  • Identifies neurobiological mechanisms based on dopamine D2-linked mechanism of action