Learn More
Medchemexpress LLC Sumatriptan-d6 succinate | 1215621-31-0 | 99.6% | 1 MG
SDP
Catalog No. 5000450795
Encompass_Preferred
Sumatriptan-d6 succinate is a deuterium-labeled version of Sumatriptan succinate. It functions as an orally active 5-HT1 receptor agonist, targeting 5-HT1D, 5-HT1B, and 5-HT1A receptors. This compound is valuable for research involving migraine headaches, where deuterium substitution can impact drug pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-50795 Supplier Medchemexpress LLC Supplier No. HYB0121BS1MG
Sumatriptan-d6 succinate is a deuterium-labeled version of Sumatriptan succinate. It functions as an orally active 5-HT1 receptor agonist, targeting 5-HT1D, 5-HT1B, and 5-HT1A receptors. This compound is valuable for research involving migraine headaches, where deuterium substitution can impact drug pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.