Learn More
Medchemexpress LLC Sumatriptan-d6 succinate | 1215621-31-0 | 99.6% | 5 MG
SDP
Catalog No. 5000450748
Encompass_Preferred
Sumatriptan-d6 succinate is a deuterium labeled form of Sumatriptan succinate. This orally active 5-HT1 receptor agonist is characterized by Kis of 17 nM for 5-HT1D, 27 nM for 5-HT1B, and 100 nM for 5-HT1A receptors, making it suitable for migraine headache research.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Orally active 5-HT1 receptor agonist
  • Deuterium labeled compound for pharmacokinetic studies
Catalog No. 50-004-50748 Supplier Medchemexpress LLC Supplier No. HYB0121BS5MG
Sumatriptan-d6 succinate is a deuterium labeled form of Sumatriptan succinate. This orally active 5-HT1 receptor agonist is characterized by Kis of 17 nM for 5-HT1D, 27 nM for 5-HT1B, and 100 nM for 5-HT1A receptors, making it suitable for migraine headache research.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Orally active 5-HT1 receptor agonist
  • Deuterium labeled compound for pharmacokinetic studies