Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sumatriptan succinate | 103628-48-4 | 99.9% | 1 ML
SDP

Catalog No. 5000450890
Encompass_Preferred

Sumatriptan succinate (GR 43175) is an orally active 5-HT1 receptor agonist that can be used for migraine headache research. It acts on 5-HT1D, 5-HT1B, and 5-HT1F receptors with IC50 values of 7.3 nM, 9.3 nM, and 17.8 nM respectively.

  • Orally active 5-HT1 receptor agonist
  • Targeting 5-HT1D, 5-HT1B, and 5-HT1F receptors
  • Suitable for migraine headache research

Catalog No. 50-004-50890 Supplier Medchemexpress LLC Supplier No. HYB012110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sumatriptan succinate (GR 43175) is an orally active 5-HT1 receptor agonist that can be used for migraine headache research. It acts on 5-HT1D, 5-HT1B, and 5-HT1F receptors with IC50 values of 7.3 nM, 9.3 nM, and 17.8 nM respectively.

  • Orally active 5-HT1 receptor agonist
  • Targeting 5-HT1D, 5-HT1B, and 5-HT1F receptors
  • Suitable for migraine headache research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.