Learn More
TARGETMOL CHEMICALS INC Sumatriptan succinate 200MG
SDP
Catalog No. 502880357
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Sumatriptan succinate (GR 43175), a serotonin1 (5-HT1) receptor agonist, is used in the acute treatment of a migraine headache. Purity 99.67%
Catalog No. 50-288-0357 Supplier TARGETMOL CHEMICALS INC Supplier No. T0203200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Sumatriptan succinate (GR 43175), a serotonin1 (5-HT1) receptor agonist, is used in the acute treatment of a migraine headache. Purity 99.67%