Learn More
Selleck Chemical LLC Sumatriptan Succinate S1432-25mg
SDP
Catalog No. 503110624
Encompass_Preferred
Sumatriptan Succinate (GR 43175) is a triptan sulfa drug containing a sulfonamide group for the treatment of migraine headaches
Catalog No. 50-311-0624 Supplier Selleck Chemical LLC Supplier No. S143225MG
Sumatriptan Succinate (GR 43175) is a triptan sulfa drug containing a sulfonamide group for the treatment of migraine headaches