Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sumatriptan Succinate, EP, 97.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18607182 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-182 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-182 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S34351KGBL

missing translation for 'validMainframeMsg'

S3435, 103628-48-4, C18H27N3O6S

Spectrum™ Chemical Sumatriptan Succinate, EP , a synthetic drug part of the triptan class, is used to treat migraine headaches. The EP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 103628-48-4
Molecular Formula C18H27N3O6S
Molecular Weight (g/mol) 413.49
InChI Key PORMUFZNYQJOEI-UHFFFAOYSA-N
IUPAC Name 1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
SMILES OC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1

Specifications

CAS Max % 1
pH 4.5 to 5.3
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 97.5 to 102%
Grade EP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.