Learn More
Aviva Systems Biology Corp SUMF1 Antibody - C-terminal region (ARP64171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUMF1. It was validated on Western Blot. . Alternate names: AAPA3037, FGE, MGC131853, MGC150436, UNQ3037ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-195