Learn More
Enzo Life Sciences SUMO-1 (93-97)-AMCA (1 mg)
SDP
Supplier:  Enzo Life Sciences BMLUW05001000
Encompass_Preferred
SUMO1-AMCA is a synthetic AMCA-peptide substrate based on the C-terminal residues of SUMO1 and is a useful fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760). It has particular utility for the study of deSUMOylating activity where continuous monitoring of activity is essential. It provides an alternative substrate to SUMO1-AMC. Purity: ≥95% (HPLC). Formulation: Lyophilized solid. Solubility: Soluble in DMSO. Can be subsequently diluted in aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-201-2250