Learn More
Enzo Life Sciences SUMO-1 (human), (recombinant) (His-tag) (500 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW91950500
Encompass_Preferred
Alternative name: Sentrin 1, Small ubiquitin related modifier 1. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In HEPES, pH 8.0, containing 50mM NaCl, 1mM DTT. Source: Produced in E. coli. Human SUMO-1 is fused to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2305