Learn More
Enzo Life Sciences SUMO-1 (human) (CT) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW94600100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 72-97 of human SUMO-1. Source: Purified from rabbit serum. Species reactivity: Human. Applications: WB. Purity: Partially purified by salt precipitation. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1689