Learn More
Enzo Life Sciences SUMO-1 (human) (NT) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW83300100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1-16 of human SUMO-1. Source: Purified from rabbit serum. Species reactivity: Human. Applications: IP, WB. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1621