Learn More
Enzo Life Sciences SUMO-1 (human) polyclonal antibody (25µg)
SDP
Supplier:  Enzo Life Sciences BMLPW0505A0025
Encompass_Preferred
Immunogen: Full length recombinant human SUMO-1. Source: Purified from rabbit serum. Species reactivity: Human. Applications: ICC, IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-1530