Learn More
Enzo Life Sciences SUMO-2/3 (human) (NT) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW94650100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1-15 of human SUMO-2. Source: Purified from rabbit serum. Species reactivity: Human. Applications: WB. Purity: Partially purified by salt precipitation. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1691