Learn More
Enzo Life Sciences SUMO-2 (human), (recombinant) (His-tag) (500 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW92050500
Encompass_Preferred
Mature protein for functional studies. Mature protein for functional studies. Alternative name: Sentrin 2, Small ubiquitin related modifier 2, SUMO-3. Formulation: Liquid. In 20mM HEPES, pH 8.0, containing 50mM NaCl, 1mM DTT. Source: Produced in E. coli. Human SUMO-2 is fused to an N-terminal His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2306