Learn More
Enzo Life Sciences SUMO-2 (human) (1-93), (recombinant) (500µg)
SDP
Supplier:  Enzo Life Sciences ALX201089C500
Encompass_Preferred
Active form of human SUMO-2 for conjugation to protein substrates. Active form of human SUMO-2 for conjugation to protein substrates. Alternative name: Sentrin 2, Small ubiquitin related modifier 2, SUMO-3. Purity: ≥95%. Formulation: Liquid. In PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Use/Stability: Stable for a few days at room temperature.  .
Catalog No. 50-200-7914