Learn More
Enzo Life Sciences SUMO-3 (human), (recombinant) (His-tag) (500 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW92150500
Encompass_Preferred
Mature protein for functional studies. Mature protein for functional studies. Alternative name: Sentrin 3, Small ubiquitin related modifier 3, SUMO-2. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM HEPES, pH 8.0, containing 50mM sodium chloride and 1mM dithiothreitol. Source: Produced in E. coli. Human SUMO-3 is fused to an N-terminal His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2307