Learn More
Enzo Life Sciences SUMO-4 (human) (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW09050100
Encompass_Preferred
Alternative name: Small ubiquitin related modifier 4. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 20mM HEPES, pH 8.0, containing 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2274