Learn More
Enzo Life Sciences SUMO activating enzyme E1 (human), (recombinant) (25 µg)
SDP
Catalog No. 502012314
Encompass_Preferred
Formulation: Liquid. In HEPES, pH 8.0, containing 110mM potassium acetate, 2mM magnesium acetate, 0.5mM EGTA and 1mM DTT. Source: Produced in E. coli. Full length heterodimeric human SUMO-activating enzyme. Long Term Storage: -80°C.
Catalog No. 50-201-2314 Supplier Enzo Life Sciences Supplier No. BMLUW93300025
Formulation: Liquid. In HEPES, pH 8.0, containing 110mM potassium acetate, 2mM magnesium acetate, 0.5mM EGTA and 1mM DTT. Source: Produced in E. coli. Full length heterodimeric human SUMO-activating enzyme. Long Term Storage: -80°C.