Learn More
LifeSensors SUMO Protease 1 250 Units
SDP
Catalog No. 501037441
Encompass_Preferred
LifeSensors' industry leading SUMO Protease 1 (Ulp1), a highly active and robust recombinant protease, cleaves SUMO from recombinant fusion proteins. Unlike thrombin, EK, or TEV protease, which recognize short linear sequences, SUMO Protease 1 recognizes the tertiary structure of SUMO. As a result, SUMO Protease 1 will not cleave within the fused protein of interest. 1 unit will cleave >90 µg of hSUMO3-GFP in 1 hr at 37C.
Catalog No. 50-103-7441 Supplier LifeSensors Supplier No. SP40100250
LifeSensors' industry leading SUMO Protease 1 (Ulp1), a highly active and robust recombinant protease, cleaves SUMO from recombinant fusion proteins. Unlike thrombin, EK, or TEV protease, which recognize short linear sequences, SUMO Protease 1 recognizes the tertiary structure of SUMO. As a result, SUMO Protease 1 will not cleave within the fused protein of interest. 1 unit will cleave >90 µg of hSUMO3-GFP in 1 hr at 37C.