Learn More
ACROBiosystems SUMO Protease, 1000 U, Remove SUMO-tag in any recombinant SUMO fusion protein.
Catalog No. NC3738124
Encompass_Preferred
SUMO Protease is a recombinant form of Saccharomyces cerevisiae protease produced in Escherichia coli. SUMO protease can recognize the SUMO tertiary structure and cleave it, has high specificity, and there would be no amino acid residues. This product contains His-tag which can be removed by Ni2+ affinity chromatography resin.
Catalog No. NC3738124 Supplier ACROBiosystems Supplier No. SUES51271000U
SUMO Protease is a recombinant form of Saccharomyces cerevisiae protease produced in Escherichia coli. SUMO protease can recognize the SUMO tertiary structure and cleave it, has high specificity, and there would be no amino acid residues. This product contains His-tag which can be removed by Ni2+ affinity chromatography resin.