Learn More
LifeSensors SUMO PROTEASE 2 500 UNITS
SDP
Catalog No. 501037447
Encompass_Preferred
For removal of SUMOpro3 tags from fusion proteins expressed from the following SUMOpro3 expression systems: (Cat.#s: 1010A, 1016, 1010K, 1011, 2151). SUMO Protease 2, a highly active and robust recombinant protease, cleaves hSUMO3 from recombinant fusion proteins. Unlike thrombin, EK, or TEV protease, which recognize short, linear sequences, SUMO Protease 2 recognizes the tertiary structure of huSUMO3. As a result, SUMO Protease 2 will not cleave within the fused protein of interest. By purchasing this product, the purchaser agrees to comply with the terms of our Limited Use Label License.
Catalog No. 50-103-7447 Supplier LifeSensors Supplier No. SP40200500
For removal of SUMOpro3 tags from fusion proteins expressed from the following SUMOpro3 expression systems: (Cat.#s: 1010A, 1016, 1010K, 1011, 2151). SUMO Protease 2, a highly active and robust recombinant protease, cleaves hSUMO3 from recombinant fusion proteins. Unlike thrombin, EK, or TEV protease, which recognize short, linear sequences, SUMO Protease 2 recognizes the tertiary structure of huSUMO3. As a result, SUMO Protease 2 will not cleave within the fused protein of interest. By purchasing this product, the purchaser agrees to comply with the terms of our Limited Use Label License.