Learn More
Genscript Corporation Antibody Enzyme Proteins Proteins 5000 UN 1/EA
Supplier:  Genscript Corporation Z036915000
Encompass
GenScripts SUMO Protease Animal-Free His is the recombinant SUMO protease fragment from Saccharomyces cerevisiae. . It is produced under an animal free process and is suitable for drug and vaccine development manufacture and other applications.
Catalog No. 50-252-9580