Learn More
AURORA BIOLABS LLC SUMO Protease (Ulp1), 1000 units
Catalog No. NC3787971
Encompass
SUMO Protease otherwise known as Ulp1, is a recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae. This highly specific protease cleaves off sumo tags by recognizing the tertiary structure of SUMO as opposed to recognizing its amino acid sequence. Because of its recognition of SUMO’s tertiary structure, it has little to no non-specific proteolysis keeping your protein samples safe after all affinity tags have been cleaved off. SUMO protease is active from 2°C to 37°C in a pH range of 7.0 – 9.0, for lower temperatures allow more time for the cleavage reaction.
Catalog No. NC3787971 Supplier AURORA BIOLABS LLC Supplier No. 1900031K
SUMO Protease otherwise known as Ulp1, is a recombinant fragment of ULP1 (Ubl-specific protease 1) from Saccharomyces cerevisiae. This highly specific protease cleaves off sumo tags by recognizing the tertiary structure of SUMO as opposed to recognizing its amino acid sequence. Because of its recognition of SUMO’s tertiary structure, it has little to no non-specific proteolysis keeping your protein samples safe after all affinity tags have been cleaved off. SUMO protease is active from 2°C to 37°C in a pH range of 7.0 – 9.0, for lower temperatures allow more time for the cleavage reaction.