Learn More
Enzo Life Sciences SUMO-QAPTURE-T kit (10 tests)
SDP
Catalog No. 502012276
Encompass_Preferred
The SUMO-QAPTURE-T kit is an efficient tool for the selective isolation of SUMOylated proteins. The kit facilitates the affinity purification of SUMOylated proteins from cell extracts and tissue lysates using a high-binding SIM-containing affinity matrix. Captured proteins are eluted under denaturing conditions followed by analysis by Western blotting, using the SUMO antibodies provided or antibodies to specific proteins of interest, or potential substrate identification by proteomic methods. SUMO-conjugate-containing samples are prepared in native form in the presence of protease inhibitors to prevent loss through the action of deSUMOylating enzymes. Optimization of binding permits complete isolation of the full range of SUMO-protein conjugates from a specific lysate.
Catalog No. 50-201-2276 Supplier Enzo Life Sciences Supplier No. BMLUW1000A0001
The SUMO-QAPTURE-T kit is an efficient tool for the selective isolation of SUMOylated proteins. The kit facilitates the affinity purification of SUMOylated proteins from cell extracts and tissue lysates using a high-binding SIM-containing affinity matrix. Captured proteins are eluted under denaturing conditions followed by analysis by Western blotting, using the SUMO antibodies provided or antibodies to specific proteins of interest, or potential substrate identification by proteomic methods. SUMO-conjugate-containing samples are prepared in native form in the presence of protease inhibitors to prevent loss through the action of deSUMOylating enzymes. Optimization of binding permits complete isolation of the full range of SUMO-protein conjugates from a specific lysate.