Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7341MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Additionally, this antibody has been validated by MS-QAVA(TM). Specificity has been quantitatively determin. Conjugate/Tag: Unconjugated. Clone: SM1/495. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Breast carcinoma. Cellular Staining: Predominantly nuclear with some cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human SUMO1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 11.5kDa (Monomer); 90kDa (Heteromer).Chromosome: 2q33.1. Gene Symbol: SUMO1. Gene ID: 7341. Ensemble: ENSG00000116030. Uniprot: NP_001005782.
Catalog No. 50-188-5261