Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6613MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SM23/496. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa cells or breast carcinoma. Cellular Staining: Nuclear (SUMO-2); Cytoplasmic (SUMO-3). Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Mouse / IgG1, kappa. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 11-13kDa.Chromosome: 17q25.1 (SUMO-2) & 21q22.3 (SUMO-3). Gene Symbol: SUMO2 & SUMO3. Gene ID: 6613. Ensemble: ENSG00000188612. Uniprot: NP_008868.
Catalog No. 50-188-5171