Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6613MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM621. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa cells or breast carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human SUMO2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 11-13kDa.Chromosome: 17q25.1. Gene Symbol: SUMO2. Gene ID: 6613. Ensemble: ENSG00000188612. Uniprot: NP_008868.
Catalog No. 50-188-5174