Learn More
Aviva Systems Biology Corp SUMO3 Antibody - middle region (ARP58247_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58247_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SUMO3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SUMO3. Alternate names: SMT3A, SMT3H1, SUMO-3, Smt3BApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-014-62269