Learn More
Aviva Systems Biology Corp Sumo3 Antibody - N-terminal region (ARP45841_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45841_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sumo3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Sumo3. Alternate names: 2810014B19Rik, AA409334, AW121497, AW536077, D10Ertd345e, SMT3A, SUMO-3, Smt3h1, Smt3BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-220