Learn More
ABclonal Technology SUMO3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15724500UL
Encompass_Preferred
This gene encodes a member of the small ubiquitin-related modifier (SUMO) family of eukaryotic proteins. The encoded protein is covalently conjugated to other proteins via a post-translation modification known as sumoylation. Sumoylation may play a role in a wide variety of cellular processes, including nuclear transport, DNA replication and repair, mitosis, transcriptional regulation, and signal transduction. Alternatively spliced transcript variants encoding distinct proteins have been described.
Catalog No. 50-001-07918