Learn More
Justrite™ Sump-to-Sump™ Drain Kit for EcoPolyBlend™ Accumulation Centers, Stainless Steel Clips, Grommets, Transfer Tube
Catalog No. 19822446 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
EcoPolyBlend Accumulation Centers
Catalog No. For Use With (Equipment)
19-822-446 EcoPolyBlend Accumulation Centers
1 options
Catalog No. 19-822-446 Supplier Justrite™ Supplier No. 28927
  • $115.00
    $77.65 / Each of 1
    Save $37.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Sump-to-Sump™ Drain Kit for EcoPolyBlend™ Accumulation Centers, Stainless Steel Clips, Grommets, Transfer Tube is an accessory to help facilitate the use of our accumulation centers.
Use for applications requiring a continuous sump and for full compliance with regulations when sump capacities reach 71 gal. (270 L). Drain kit includes one pair stainless-steel joining clips, one pair rubber grommets and polytetrafluoroethylene transfer tube. It allows you to join centers together, on site, using unites 4-drum and smaller, to reach needed capacity. By joining centers together with the patented Sump-to-Sump Drain™ kit for a total 66-gallon sump capacity, you will meet EPA and UFC requirements.
Specifications
For Use With (Equipment) EcoPolyBlend Accumulation Centers